missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human ATPase Na+/K+ beta 2 His Protein

Product Code. 18358455 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 μg
100 μg
20 μg
250 μg
50 μg
500 μg
Unit Size:
100µg
10µg
20µg
250µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18358455 500 μg 500µg
18366976 50 μg 50µg
18310917 250 μg 250µg
18399804 20 μg 20µg
18304633 10 μg 10µg
18383782 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 18358455 Supplier Novus Biologicals™ Supplier No. NBP315116500UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Specific activity is > 3,000 pmol/min/ug, and is defined as the amount of enzyme that hydrolyze 1.0 pmole of Adenosine 5-triphosphate to phosphate per minute per minute at pH 7.5 at 25C.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.5 mg/ml
For Use With (Application) Bioactivity, SDS-PAGE
Formulation PBS (pH 7.4), 10% glycerol
Gene ID (Entrez) 482
Molecular Weight (g/mol) M.W. Theoretical: 26.4 kDa
Quantity 500 μg
Storage Requirements Store at 4C short term. Aliquot and Store at -20°C long term. Avoid freeze-thaw cycles.
Endotoxin Concentration <1 EU per 1ug of protein (determined by LAL method)
Gene Alias adhesion molecule on glia, AMOGsodium/potassium-dependent ATPase beta-2 subunit, ATPase, Na+/K+ transporting, beta 2 polypeptide, Na, K-ATPase beta-2 polypeptide, Sodium/potassium-dependent ATPase subunit beta-2, sodium/potassium-transporting ATPase beta-2 chain, sodium/potassium-transporting ATPase subunit beta-2
Gene Symbol ATP1B2
Research Category Endocrinology, Signal Transduction
Purity or Quality Grade >90%, by SDS-PAGE
Protein ATPase Na+/K+ beta 2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.