missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ATP5J, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) ATP5J, mitochondrial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5697-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001003696.1 | |
ATP5J, mitochondrial Protein | |
Research Use Only | |
ATP5J | |
Recombinant Protein | |
E. coli | |
NKELDPIQKL FVDKIREYKS KRQTSGGPVD ASSEYQQELE RELFKLKQMF GNADMNTFPT FKFEDPKFEV IEKPQA | |
Greater than 80% as determined by SDS-PAGE. |
522 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |