missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ATP5J, mitochondrial Protein

A DNA sequence encoding the Homo sapiens (Human) ATP5J, mitochondrial, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5697-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15947089

  • £560.00 / 100µg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001003696.1
ATP5J, mitochondrial Protein
Research Use Only
ATP5J
Recombinant Protein
E. coli
NKELDPIQKL FVDKIREYKS KRQTSGGPVD ASSEYQQELE RELFKLKQMF GNADMNTFPT FKFEDPKFEV IEKPQA
Greater than 80% as determined by SDS-PAGE.
522
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers