missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ATP5D Protein
A DNA sequence encoding the Homo sapiens (Human) ATP5D, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8328-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001001975.1 | |
ATP5D Protein | |
Research Use Only | |
ATP5D | |
Recombinant Protein | |
E. coli | |
ASPTQVFFNG ANVRQVDVPT LTGAFGILAA HVPTLQVLRP GLVVVHAEDG TTSKYFVSSG SIAVNADSSV QLLAEEAVTL DMLDLGAAKA NLEKAQAELV GTADEATRAE IQIRIEANE | |
Greater than 87.9% as determined by SDS-PAGE. |
513 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |