missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ATP5D Protein

A DNA sequence encoding the Homo sapiens (Human) ATP5D, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8328-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16004041

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001001975.1
ATP5D Protein
Research Use Only
ATP5D
Recombinant Protein
E. coli
ASPTQVFFNG ANVRQVDVPT LTGAFGILAA HVPTLQVLRP GLVVVHAEDG TTSKYFVSSG SIAVNADSSV QLLAEEAVTL DMLDLGAAKA NLEKAQAELV GTADEATRAE IQIRIEANE
Greater than 87.9% as determined by SDS-PAGE.
513
200 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers