missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ATP-dependent RNA helicase A Protein

A DNA sequence encoding the Homo sapiens (Human) ATP-dependent RNA helicase A, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7512-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16039110

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001348.2
ATP-dependent RNA helicase A Protein
Research Use Only
DHX9
Recombinant Protein
E. coli
NQVGVVPWSP PQSNWNPWTS SNIDEGPLAF ATPEQISMDL KNELMYQLEQ DHDLQAILQE RELLPVKKFE SEILEAISQN SVVIIRGATG CGKTTQVPQF ILDDFIQNDR AAECNIVVTQ PRRISAVSVA ERVAFERGEE PGKSCGYSVR FESILPRPHA SIMFCTVGVL LRKLEAGIRG ISHVIVDEIH ERDINTDFLL VVLRDVVQAY PEVRIVLMSA TIDTSMFCEY FFNCPIIEVY GRTYPVQEYF LEDCIQMTHF VPPPKDKKKK DKDDDGGEDD DANCNLICGD EYGPETRLSM SQLNEKETPF ELIEALLKYI ETLNVPGAVL VFLPGWNLIY TMQKHLEMNP HFGSHRYQIL PLHSQIPREE QRKVFDPVPV GVTKVILSTN IAETSITIND VVYVIDSCKQ KVKLFTAHNN MTNYATVWAS KTNLEQRKGR AGRVRPGFCF HLCSRARFER LETHMTPEMF RTPLHEIALS IKLLRLGGIG QFLAKAIEPP PLDAVIEAEH TLRELD
Greater than 90% as determined by SDS-PAGE.
1660
10 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers