missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ATF Protein
A cDNA sequence encoding the ATF was constructed and used to recombinantly synthesize the protein.
£283.00 - £1765.00
Specifications
Gene ID (Entrez) | 7018 |
---|---|
Name | ATF Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | TF |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15946128
|
enQuireBio™
QP11096-500MG |
500 mg |
£283.00
500mg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15936128
|
enQuireBio™
QP11096-1GRAM |
1 g |
£439.00
1g |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15956128
|
enQuireBio™
QP11096-5GRAM |
5 g |
£1765.00
5g |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
7018 | |
Research Use Only | |
TF | |
Human | |
Untagged | |
The protein (10 mg/ml) was lyophilized from 20mM NH4HC03 solution. May contain traces of buffer salts. |
ATF Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Human | |
MRLAVGALLV CAVLGLCLAV PDKTVRWCAV SEHEATKCQS FRDHMKSVIP SDGPSVACVK KASYLDCIRA IAANEADAVT LDAGLVYDAY LAPNNLKPVV AEFYGSKEDP QTFYYAVAVV KKDSGFQMNQ LRGKKSCHTG LGRSAGWNIP IGLLYCDLPE PRKPLEKAVA NFFSGSCAPC ADGTDFPQLC QLCPGCGCST LNQYFGYSGA FKCLKDGAGD VAFVKHSTIF ENLANKADRD QYELLCLDNT RKPVDEYKDC HLAQVPSHTV VARSMGGKED LIWELLNQAQ EHFGKDKSKE FQLFSSPHGK DLLFKDSAHG FLKVPPRMDA KMYLGYEYVT AIRNLREGTC PEAPTDECKP VKWCALSHHE RLKCDEWSVN SVGKIECVSA ETTEDCIAKI MNGEADAMSL DGGFVYIAGK CGLVPVLAEN YNKSDNCEDT PEAGYFAIAV VKKSASDLTW DNLKGKKSCH TAVGRTAGWN IPMGLLYNKI NHCRFDEFFS EGCAPGSKKD SSLCKLCMGS GLNLCEPNNK EGYYGYTGAF RCLVEKGDVA FVKHQTVPQN TGGKNPDPWA KNLNEKDYEL LCLDGTRKPV EEYANCHLAR APNHAVVTRK DKEACVHKIL RQQQHLFGSN VTDCSGNFCL FRSETKDLLF RDDTVCLAKL HDRNTYEKYL GEEYVKAVGN LRKCSTSSLL EACTFRRP | |
Greater than 98.0% as determined by coomassie blue stained SDS-PAGE and Cellulose Acetate electrophoresis. |