missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Astrocytic phosphoprotein PEA-15 Protein

A DNA sequence encoding the Homo sapiens (Human) Astrocytic phosphoprotein PEA-15, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7581-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16059540

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001284505.1
Astrocytic phosphoprotein PEA-15 Protein
Research Use Only
PEA15
Recombinant Protein
E. coli
MAEYGTLLQD LTNNITLEDL EQLKSACKED IPSEKSEEIT TGSAWFSFLE SHNKLDKDNL SYIEHIFEIS RRPDLLTMVV DYRTRVLKIS EEDELDTKLT RIPSAKKYKD IIRQPSEEEI IKLAPPPKKA
Greater than 80% as determined by SDS-PAGE.
8682
10 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers