missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Astrocytic phosphoprotein PEA-15 Protein

A DNA sequence encoding the Homo sapiens (Human) Astrocytic phosphoprotein PEA-15, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7581-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16049540

  • £384.00 / 100µg
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001284505.1
Astrocytic phosphoprotein PEA-15 Protein
Research Use Only
PEA15
Recombinant Protein
E. coli
MAEYGTLLQD LTNNITLEDL EQLKSACKED IPSEKSEEIT TGSAWFSFLE SHNKLDKDNL SYIEHIFEIS RRPDLLTMVV DYRTRVLKIS EEDELDTKLT RIPSAKKYKD IIRQPSEEEI IKLAPPPKKA
Greater than 80% as determined by SDS-PAGE.
8682
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers