missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Asparagine Synthetase/ASNS His Protein
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
Description
- Although the name focuses attention on its function in asparagine synthesis, the reaction catalyzed by ASNS also impacts glutamine, aspartate and glutamate homeostasis.
- Although ubiquitously expressed at low levels in many organs, expression is particularly high in the pancreas.
Specifications
Specifications
| Accession Number | P08243-1 |
| For Use With (Application) | Enzyme Activity |
| Gene ID (Entrez) | 440 |
| Molecular Weight (g/mol) | 65 kDa |
| Purification Method | >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining |
| Quantity | 20 μg |
| Source | Human embryonic kidney cell, HEK293-derived human Asparagine Synthetase/ASNS protein Cys2-Ala561 with C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70°C as supplied. 3 months, -20 to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method |
| Conjugate | Unconjugated |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction