missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human ASIP/Agouti Protein

Product Code. 18745117
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18745117

Brand: R&D Systems™ 9094AG050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human ASIP/Agouti Protein is derived from HEK293. The Recombinant Human ASIP/Agouti Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P42127
For Use With (Application) Bioactivity
Gene ID (Entrez) 434
Molecular Weight (g/mol) Observed M.W.: 18-23kDa, reducing conditions, Theoretical M.W.: 12kDa
Quantity 50 μg
Source Human embryonic kidney cell,HEK293-derived human ASIP protein His23-Cys132
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias agouti (mouse)-signaling protein, agouti signaling protein, agouti signaling protein, nonagouti homolog, agouti signaling protein, nonagouti homolog (mouse), Agouti switch protein, agouti-signaling protein, AGSW, AGTI, AGTIL, AGTILMGC126092, ASPAGSW, MGC126093, nonagouti homolog, SHEP9
Common Name ASIP
Gene Symbol ASIP
Biological Activity Measured by its ability to inhibit alpha-MSH induced eumelanin production in B16F1 mouse melanoma cells. The ED50 for this effect is typically 0.8-4μg/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Human embryonic kidney cell, HEK293-derived His23-Cys132
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.