missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human AS3MT Protein

Product Code. 18178070
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18178070

Brand: R&D Systems™ 9827MT050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human AS3MT Protein is derived from E. coli. The Recombinant Human AS3MT Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9HBK9-1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in Tris and NaCl with Trehalose.
Gene ID (Entrez) 57412
Molecular Weight (g/mol) 42 kDa
Name arsenic (+3 oxidation state) methyltransferase, Arsenic Methyltransferase, arsenite methyltransferase, CYT19, CYT19methyltransferase cyt19, EC 2.1.1.137, Methylarsonite methyltransferase, Methyltransferase Cyt19, S-Adenosyl-L-Methionine:Arsenic(III) Methyltransfe, S-adenosyl-L-methionine:arsenic(III) methyltransferase, S-Adenosylmethionine:Arsenic (III) Methyltransfera, S-adenosylmethionine:arsenic (III) methyltransferase
Quantity 50 μg
Source E. coli-derived human AS3MT protein Ala2-Cys375,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 500μg/mL in water.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.