missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Artemin / ARTN Protein

Product Code. 15975919
Click to view available options
Quantity:
1 mg
100 μg
20 μg
5 μg
500 μg
Unit Size:
100µg
1mg
20µg
500µg
5µg
This item is not returnable. View return policy

Product Code. 15975919

Brand: enQuireBio™ QP53305ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Specifications

Accession Number NP_001129687.1
Gene ID (Entrez) 9048
Name Artemin / ARTN Protein
Quantity 5 μg
Regulatory Status Research Use Only
Endotoxin Concentration Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Gene Symbol Artn
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag Untagged
Sequence AGGPGSRARA AGARGCRLRS QLVPVRALGL GHRSDELVRF RFCSGSCRRA RSPHDLSLAS LLGAGALRPP PGSRPVSQPC CRPTRYEAVS FMDVNSTWRT VDRLSATACG CLG
Buffer 10 mM sodium citrate, 25 mM sodium chloride, pH 4.5
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.