missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ARL4D Protein
A cDNA sequence encoding the ARL4D was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11072-1mg
Additional Details : Weight : 0.01000kg
Specifications
ARL4D Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGNHLTEMAP TASSFLPHFQ ALHVVVIGLD SAGKTSLLYR LKFKEFVQSV PTKGFNTEKI RVPLGGSRGI TFQVWDVGGQ EKLRPLWRSY TRRTDGLVFV VDAAEAERLE EAKVELHRIS RASDNQGVPV LVLANKQDQP GALSAAEVEK RLAVRELAAA TLTHVQGCSA VDGLGLQQGL ERLYEMILKR KKAARGGKKR R | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The ARL4D solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 2mM DTT, 0.2M NaCl and 20% glycerol. |