missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ARL1 Protein
A cDNA sequence encoding the ARL1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11067-1mg
Additional Details : Weight : 0.01000kg
Specifications
ARL1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGGFFSSIFS SLFGTREMRI LILGLDGAGK TTILYRLQVG EVVTTIPTIG FNVETVTYKN LKFQVWDLGG QTSIRPYWRC YYSNTDAVIY VVDSCDRDRI GISKSELVAM LEEEELRKAI LVVFANKQDM EQAMTSSEMA NSLGLPALKD RKWQIFKTSA TKGTGLDEAM EWLVETLKSR Q | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The ARL1 solution (0.25 mg/ml) containing 20mM Tris-HCl pH 8.0, 2mM DTT, 100mM NaCl and 40% glycerol. |