missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ARHGEF39 Protein

A cDNA sequence encoding the ARHGEF39 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11066-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15956038

  • £99.15 / 5µg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

ARHGEF39 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMELSCPG SRCPVQEQRA RWERKRACTA RELLETERRY QEQLGLVATY FLGILKAKGT LRPPERQALF GSWELIYGAS QELLPYLEGG CWGQGLEGFC RHLELYNQFA ANSERSQTTL QEQLKKNKGF RRFVRLQEGR PEFGGLQLQD LLPLPLQRLQ QYENLVVALA ENTGPNSPDH QQLTRAARLI SETAQRVHTI GQKQKNDQHL RRVQALLSGR QAKGLTSGRW FLRQGWLLVV PPHGEPRPRM FFLFTDVLLM AKPRPPLHLL RSGTFACKAL YPMAQCHLSR VFGHSGGPCG GLLSLSFPHE KLLLMSTDQE ELSRWYHSLT WAISSQKN
Greater than 85.0% as determined by SDS-PAGE.
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
ARHGEF39 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers