missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human ARHGEF19 GST (N-Term) Protein

Product Code. 18615131 Shop All Bio Techne Products
Click to view available options
Quantity:
2 μg
Unit Size:
2µg
This item is not returnable. View return policy

Product Code. 18615131

Brand: Novus Biologicals™ H00128272P012ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Recombinant Protein

This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blot, ELISA, Protein Array, Immunoaffinity Purification
Formulation 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
Gene ID (Entrez) 128272
Molecular Weight (g/mol) MolecularWeight-theroretical: 115.6 kDa
Quantity 2 μg
Storage Requirements Store at -80°C. Avoid freeze/thaw cycles.
Gene Alias ephexin-2, FLJ33962, Rho guanine nucleotide exchange factor (GEF) 19, RP4-733M16.1, weakly similar to Rho GEF, WGEFrho guanine nucleotide exchange factor 19
Gene Symbol ARHGEF19
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein ARHGEF19
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.