missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human APTX Protein

A cDNA sequence encoding the APTX was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11059-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15946018

  • £99.15 / 2µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

APTX Protein
Research Use Only
Recombinant Protein
E. coli
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWAGSMQD PKMQVYKDEQ VVVIKDKYPK ARYHWLVLPW TSISSLKAVA REHLELLKHM HTVGEKVIVD FAGSSKLRFR LGYHAIPSMS HVHLHVISQD FDSPCLKNKK HWNSFNTEYF LESQAVIEMV QEAGRVTVRD GMPELLKLPL RCHECQQLLP SIPQLKEHLR KHWTQ
Greater than 95.0% as determined by SDS-PAGE.
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The APTX solution contains 20mM Tris-HCl pH 7.5, 0.1M NaCl, 0.1mM PMSF and 20% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers