missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human Apolipoprotein A-I/ApoA1 Protein

Product Code. 18462092 Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
100 μg
500 μg
Unit Size:
100µg
1mg
500µg
This item is not returnable. View return policy

Product Code. 18462092

Brand: Novus Biologicals™ NBP234869500ug

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Recombinant Protein

Specifications

Accession Number P02647, P02647, P02647
Concentration LYOPH
For Use With (Application) PAGE
Formulation Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4.
Gene ID (Entrez) 335
Molecular Weight (g/mol) 28.1 kDa
Quantity 500 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles.
Endotoxin Concentration Less than 0.1 EU/ug of Apolipoprotein A-I/ApoA1 as determined by LAL method.
Gene Alias APOA1, Apo-AI, ApoA-I, Apolipoprotein A1, apolipoprotein A-I, MGC117399
Gene Symbol APOA1
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C.
Research Category Biologically Active Proteins, Breast Cancer, Cancer, Diabetes Research, Lipid and Metabolism
Purity or Quality Grade >98% pure by SDS-PAGE and HPLC
Protein Apolipoprotein A-I/ApoA1
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.