missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human AP1S2 Protein

A cDNA sequence encoding the AP1S2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11039-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15945948

  • £151.37 / 10µg
Estimated Shipment: 22-02-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

AP1S2 Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
AP1S2 protein solution (0.5 mg/1ml) containing 20mM Tris-HCl buffer (pH 8.0), 40% glycerol, 0.1M NaCl and 2mM DTT.
Greater than 85.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MQFMLLFSRQ GKLRLQKWYV PLSDKEKKKI TRELVQTVLA RKPKMCSFLE WRDLKIVYKR YASLYFCCAI EDQDNELITL EIIHRYVELL DKYFGSVCEL DIIFNFEKAY FILDEFLLGG EVQETSKKNV LKAIEQADLL QEEAETPRSV LEEIGLT
Videos
SDS
Documents
Certificates
Special Offers

Special Offers