missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ANXA13 Protein

A cDNA sequence encoding the ANXA13 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11033-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15975928

  • £155.00 / 20µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

ANXA13 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGNRHAKASS PQGFDVDRDA KKLNKACKGM GTNEAAIIEI LSGRTSDERQ QIKQKYKATY GKELEEVLKS ELSGNFEKTA LALLDRPSEY AARQLQKAMK GLGTDESVLI EVLCTRTNKE IIAIKEAYQR LFDRSLESDV KGDTSGNLKK ILVSLLQANR NEGDDVDKDL AGQDAKDLYD AGEGRWGTDE LAFNEVLAKR SYKQLRATFQ AYQILIGKDI EEAIEEETSG DLQKAYLTLV RCAQDCEDYF AERLYKSMKG AGTDEETLIR IVVTRAEVDL QGIKAKFQEK YQKSLSDMVR SDTSGDFRKL LVALLH
Greater than 90.0% as determined by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The ANXA13 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 30% glycerol, 0.1M NaCl and 1mM EDTA.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers