missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ANP32A Protein
A cDNA sequence encoding the ANP32A was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11026-1mg
Additional Details : Weight : 0.01000kg
Specifications
ANP32A Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MEMGRRIHLE LRNRTPSDVK ELVLDNSRSN EGKLEGLTDE FEELEFLSTI NVGLTSIANL PKLNKLKKLE LSDNRVSGGL EVLAEKCPNL THLNLSGNKI KDLSTIEPLK KLENLKSLDL FNCEVTNLND YRENVFKLLP QLTYLDGYDR DDKEAPDSDA EGYVEGLDDE EEDEDEEEYD EDAQVVEDEE DEDEEEEGEE EDVSGEEEED EEGYNDGEVD DEEDEEELGE EERGQKRKRE PEDEGEDDD | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
ANP32A solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 100mM NaCl, 0.1mM PMSF and 10% glycerol. |