missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ANKRD54 Protein
A cDNA sequence encoding the ANKRD54 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11025-100ug
Additional Details : Weight : 0.01000kg
Specifications
129138 | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAAAAGD ADDEPRSGHS SSEGECAVAP EPLTDAEGLF SFADFGSALG GGGAGLSGRA SGGAQSPLRY LHVLWQQDAE PRDELRCKIP AGRLRRAARP HRRLGPTGKE VHALKRLRDS ANANDVETVQ QLLEDGADPC AADDKGRTAL HFASCNGNDQ IVQLLLDHGA DPNQRDGLGN TPLHLAACTN HVPVITTLLR GGARVDALDR AGRTPLHLAK SKLNILQEGH AQCLEAVRLE VKQIIHMLRE YLERLGQHEQ RERLDDLCTR LQMTSTKEQV DEVTDLLASF TSLSLQMQSM EKR | |
Greater than 85.0% as determined by SDS-PAGE. |
ANKRD54 Protein | |
Research Use Only | |
ANKRD54 | |
Human | |
His | |
ANKRD54 protein solution (0.5 mg/ml) contains 20mM Tris-HCl buffer, (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |