missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ANKRA2 Protein

A cDNA sequence encoding the ANKRA2 was constructed and used to recombinantly synthesize the protein.

£98.39 - £1324.47

Specifications

Name ANKRA2 Protein
Product Type Recombinant Protein
Protein Tag His
Host Species E. coli
Cross Reactivity Human
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15985898
View Documents Promotion Details
enQuireBio™
QP11023-2UG
2 μg
£98.39
2µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-02-2022
Please to purchase this item. Need a web account? Register with us today!
15975898
View Documents Promotion Details
enQuireBio™
QP11023-10UG
10 μg
£151.37
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-02-2022
Please to purchase this item. Need a web account? Register with us today!
15965898
View Documents Promotion Details
enQuireBio™
QP11023-100UG
100 μg
£1324.47
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-02-2022
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

ANKRA2 Protein
His
Human
Greater than 85.0% as determined by SDS-PAGE.
ANKRA2
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMDTSTNL DIGAQLIVEE CPSTYSLTGM PDIKIEHPLD PNSEEGSAQG VAMGMKFILP NRFDMNVCSR FVKSLNEEDS KNIQDQVNSD LEVASVLFKA ECNIHTSPSP GIQVRHVYTP STTKHFSPIK QSTTLTNKHR GNEVSTTPLL ANSLSVHQLA AQGEMLYLAT RIEQENVINH TDEEGFTPLM WAAAHGQIAV VEFLLQNGAD PQLLGKGRES ALSLACSKGY TDIVKMLLDC GVDVNEYDWN GGTPLLYAVH GNHVKCVKML LESGADPTIE TDSGYNSMDL AVALGYRSVQ QVIESHLLKL LQNIKE
57763
SDS
Product Certifications