missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ANGPTL6 Protein

Product Code. 15925898
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
This item is not returnable. View return policy

Product Code. 15925898

Brand: enQuireBio™ QP110212ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the ANGPTL6 was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 83854
Name ANGPTL6 Protein
Quantity 2 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Gene Symbol ANGPTL6
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence MKHHHHHHAS RAGAPRCTYT FVLPPQKFTG AVCWSGPAST RATPEAANAS ELAALRMRVG RHEELLRELQ RLAAADGAVA GEVRALRKES RGLSARLGQL RAQLQHEAGP GAGPGADLGA EPAAALALLG ERVLNASAEA QRAAARFHQL DVKFRELAQL VTQQSSLIAR LERLCPGGAG GQQQVLPPPP LVPVVPVRLV GSTSDTSRML DPAPEPQRDQ TQRQQEPMAS PMPAGHPAVP TKPVGPWQDC AEARQAGHEQ SGVYELRVGR HVVSVWCEQQ LEGGGWTVIQ RRQDGSVNFF TTWQHYKAGF GRPDGEYWLG LEPVYQLTSR GDHELLVLLE DWGGRGARAH YDGFSLEPES DHYRLRLGQY HGDAGDSLSW HNDKPFSTVD RDRDSYSGNC ALYQRGGWWY HACAHSNLNG VWHHGGHYRS RYQDGVYWAE FRGGAYSLRK AAMLIRPLKL
Buffer Filtered (0.4 microm) and lyophilized from a 0.5 mg/ml solution containing 0.05M Acetate buffer pH 4.0.
Purity or Quality Grade Greater than 90.0% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.