missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Angiostatin Fc Chimera Protein

Product Code. 18321418
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18321418 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18321418 Supplier R&D Systems™ Supplier No. 926AS050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by the ability of the immobilized protein to support the adhesion of THP-1 human acute monocytic leukemia cells. Chavakis, T. et al. (2005) Blood 105:1036. The ED50 for this effect is 0.5-2.5 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P00747
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 5340
Molecular Weight (g/mol) M.W. (Observed): 75-85 kDa, reducing conditions; M.W. (theoretical): 67.7 kDa (monomer)
Name Angiostatin Fc Chimera
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived human Angiostatin protein Met-Asp Human IgG1 (Pro100-Lys330) IEGR Human Angiostatin (Kringle 1-4) (Val98-Ala459) (N-terminus) Accession # P00747 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Symbol PLG
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 200 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE with silver staining, under reducing conditions
Protein Angiostatin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.