missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Angiopoietin-like 3 Protein
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
The Recombinant Human Angiopoietin-like 3 Protein is derived from Sf 21 (baculovirus). The Recombinant Human Angiopoietin-like 3 Protein has been validated for the following applications: Bioactivity, Inhibition Activity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS, NaCl and CHAPS. |
| Gene ID (Entrez) | 27329 |
| Molecular Weight (g/mol) | 53.2 kDa |
| Name | Angiopoietin-like Protein 3/ANGPTL3 |
| Quantity | 50 μg |
| Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived human Angiopoietin-like Protein 3/ANGPTL3 protein Ser17-Glu460,with a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Oligomer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction