missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Angiopoietin-1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£332.60
Specifications
Gene ID (Entrez) | 284 |
---|---|
Molecular Weight (g/mol) | 56 kDa |
Name | Angiopoietin-1 |
Quantity | 25μg |
Source | Mouse myeloma cell line,NS0-derived human Angiopoietin-1 protein Ser20-Phe498,with a C-terminal 6-His tag |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18387407
|
R&D Systems™
923-AN-025 |
£332.60
25µg |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18177620
|
R&D Systems™
923-AN-025/CF |
Lyophilized from a 0.2μm filtered solution in Tris-Citrate and NaCl. |
£332.60
25µg |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Human Angiopoietin-1 Protein is derived from NS0. The Recombinant Human Angiopoietin-1 Protein has been validated for the following applications: Bioactivity.Specifications
284 | |
Angiopoietin-1 | |
Mouse myeloma cell line,NS0-derived human Angiopoietin-1 protein Ser20-Phe498,with a C-terminal 6-His tag | |
Oligomer | |
Recombinant |
56 kDa | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |