missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Aminopeptidase LRAP/ERAP2 Protein

Product Code. 18399037
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18399037 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18399037 Supplier R&D Systems™ Supplier No. 3830ZN010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to cleave the fluorogenic peptide substrate, Arg-7-amido-4-methylcoumarin (Arg-AMC). The specific activity is >50 pmol/min/μg, as measured under the described conditions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6P179
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl and Glycerol.
Gene ID (Entrez) 64167
Molecular Weight (g/mol) M.W. (Observed): 122 kDa, reducing conditions; M.W. (theoretical): 105 kDa
Name Aminopeptidase LRAP/ERAP2
Quantity 10 μg
Source Mouse myeloma cell line, NS0-derived human Aminopeptidase LRAP/ERAP2 protein Ala56-Thr960, with an N-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol ERAP2
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Aminopeptidase LRAP/ERAP2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.