missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Amino-terminal enhancer of split Protein
A DNA sequence encoding the Homo sapiens (Human) Amino-terminal enhancer of split, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5618-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001121.2 | |
Amino-terminal enhancer of split Protein | |
Research Use Only | |
AES | |
Recombinant Protein | |
E. coli | |
MCHKNGFPQE GGITAAFLQK RKLRLSKNHR PARAKVTEHV RGTRPGRATA GPAASTRAAG SLFFDRWGNR GPAGCRGSSH LPQQLKFTTS DSCDRIKDEF QLLQAQYHSL KLECDKLASE KSEMQRHYVM YYEMSYGLNI EMHKQAEIVK RLNGICAQVL PYLSQEHQQQ VLGAIERAKQ VTAPELNSII RQQLQAHQLS QLQALALPLT PLPVGLQPPS LPAVSAGTGL LSLSALGSQA HLSKEDKNGH DGDTHQEDDG EKSD | |
Greater than 80% as determined by SDS-PAGE. |
166 | |
500 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |