missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human AMBP Protein

A cDNA sequence encoding the AMBP was constructed and used to recombinantly synthesize the protein.

£98.39 - £5741.17

Specifications

Name AMBP Protein
Product Type Recombinant Protein
Protein Tag His
Host Species E. coli
Cross Reactivity Human
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15945858
View Documents Promotion Details
enQuireBio™
QP11009-HIS-2UG
2 μg
£98.39
2µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-02-2022
Please to purchase this item. Need a web account? Register with us today!
15925858
View Documents Promotion Details
enQuireBio™
QP11009-HIS-10UG
10 μg
£151.37
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-02-2022
Please to purchase this item. Need a web account? Register with us today!
15935858
View Documents Promotion Details
enQuireBio™
QP11009-HIS-1MG
1 mg
£5741.17
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 24-02-2022
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

AMBP Protein
His
Human
Greater than 96.0%.
Recombinant Protein
E. coli
MRSLGALLLL LSACLAVSAG PVPTPPDNIQ VQENFNISRI YGKWYNLAIG STCPWLKKIM DRMTVSTLVL GEGATEAEIS MTSTRWRKGV CEETSGAYEK TDTDGKFLYH KSKWNITMES YVVHTNYDEY AIFLTKKFSR HHGPTITAKL YGRAPQLRET LLQDFRVVAQ GVGIPEDSIF TMADRGECVP GEQEPEPILI PRVRRAVLPQ EEEGSGGGQL VTEVTKKEDS CQLGYSAGPC MGMTSRYFYN GTSMACETFQ YGGCMGNGNN FVTEKECLQT CRTVAACNLP IVRGPCRAFI QLWAFDAVKG KCVLFPYGGC QGNGNKFYSE KECREYCGVP GDGDEELLRF SN
SDS
Product Certifications