missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human alpha-Synuclein Monomer Protein (Biologically Active)

Product Code. 18281231 Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
200 μg
500 μg
Unit Size:
1 set
200µg
500µg
This item is not returnable. View return policy

Product Code. 18281231

Brand: Novus Biologicals™ NBP254788100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified and high bioactivity. Generating reliable and reproducible results.

An un-tagged full length Human biologically active Alpha-Synuclein recombinant protein monomer, NCBI Accession #:NP_000336.1. The Recombinant Human alpha-Synuclein Monomer Protein is derived from E. coli. The Recombinant Human alpha-Synuclein Monomer Protein has been validated for the following applications: Western Blot, In vitro assay, SDS-Page, Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) In vitro Assay, In vivo Assay, SDS-PAGE, Western Blot
Formulation PBS
Gene ID (Entrez) 6622
Name Human alpha-Synuclein Monomer Protein
Purification Method >95% pure by SDS-PAGE
Quantity 100 μg
Source E.Coli
Immunogen MDVFMKGLSKAKEGVVAAAEKTKQGVAEAAGKTKEGVLYVGSKTKEGVVHGVATVAEKTKEQVTNVGGAVVTGVTAVAQKTVEGAGSIAAATGFVKKDQLGKNEEGAPQEGILEDMPVDPDNEAYEMPSEEGYQDYEPEA
Storage Requirements Store at −80°C in the dark. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Alias alpha-Synuclein, Lewy body 4, MGC110988, non A-beta component of AD amyloid, Non-A beta component of AD amyloid, non-A4 component of amyloid, Non-A4 component of amyloid precursor, PARK1, PARK4, synuclein, alpha (non A4 component of amyloid precursor)
Gene Symbol SNCA
Biological Activity 100μM alpha synuclein protein monomer seeded with 10nM alpha synuclein protein aggregate in 25μM Thioflavin T (PBS pH 7.4, 100μL reaction volume) generated a fluorescence intensity of 13,000 Relative Fluorescence Units after incubation at 37°C with shaking at 600 rpm for 24 hours. Fluorescence was measured by excitation at 450nm and emission at 485nm on a Molecular Devices Gemini XPS microplate reader.
Product Type Recombinant Protein
Conjugate Unconjugated
Cross Reactivity Human
Recombinant Recombinant
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.