missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human alpha-L-Iduronidase/IDUA Avi Protein, CF
Click to view available options
:
10 μg
Description
Measured by its binding ability in a functional ELISA. Recombinant Human alpha-L-Iduronidase/IDUA Fc Chimera Avi-tag binds to Human alpha -L-Iduronidase/IDUA Antibody (Catalog # AF4119 ) with an ED50 of 2.00-20.0 ng/mL. Measured by its ability to cleave a fluorogenic substrate, 4-Methylumbelliferyl alpha -L-iduronide. The specific activity is >4500 pmol/min/μg, as measured under the described conditions.
Specifications
Specifications
| For Use With (Application) | Bioactivity, Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Sodium Acetate, NaCl and Glycerol. |
| Gene ID (Entrez) | 3425 |
| Molecular Weight (g/mol) | M.W.-theoretical: 98 kDa, under reducing conditions, M.W.-observed: 104-115 kDa, under reducing conditions |
| Quantity | 10 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human alpha-L-Iduronidase/IDUA protein Human IDUA (Ala26-Pro653) (N-terminus) Accession # P35475.2 Human IgG1 (Pro100-Lys330) Avi-tag (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70 °C as supplied. 3 months, -20 to -70 °C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | alphaLIduronidase, IDA, IDUA, MPS1, MPSI |
| Conjugate | Biotin |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction