missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Aldo-keto reductase family 1 member B10 Protein
A DNA sequence encoding the Homo sapiens (Human) Aldo-keto reductase family 1 member B10, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5636-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_064695.3 | |
Aldo-keto reductase family 1 member B10 Protein | |
Research Use Only | |
AKR1B10 | |
Recombinant Protein | |
E. coli | |
MATFVELSTK AKMPIVGLGT WKSPLGKVKE AVKVAIDAGY RHIDCAYVYQ NEHEVGEAIQ EKIQEKAVKR EDLFIVSKLW PTFFERPLVR KAFEKTLKDL KLSYLDVYLI HWPQGFKSGD DLFPKDDKGN AIGGKATFLD AWEAMEELVD EGLVKALGVS NFSHFQIEKL LNKPGLKYKP VTNQVECHPY LTQEKLIQYC HSKGITVTAY SPLGSPDRPW AKPEDPSLLE DPKIKEIAAK HKKTAAQVLI RFHIQRNVIV IPKSVTPARI VENIQVFDFK LSDEEMATIL SFNRNWRACN VLQSSHLEDY PFNAEY | |
Greater than 80% as determined by SDS-PAGE. |
57016 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |