missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ALDH3A1 Protein
A cDNA sequence encoding the ALDH3A1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10516-10ug
Additional Details : Weight : 0.01000kg
Specifications
218 | |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSKISEAVKR ARAAFSSGRT RPLQFRIQQL EALQRLIQEQ EQELVGALAA DLHKNEWNAY YEEVVYVLEE IEYMIQKLPE WAADEPVEKT PQTQQDELYI HSEPLGVVLV IGTWNYPFNL TIQPMVGAIA AGNAVVLKPS ELSENMASLL ATIIPQYLDK DLYPVINGGV PETTELLKER FDHILYTGST GVGKIIMTAA AKHLTPVTLE LGGKSPCYVD KNCDLDVACR RIAWGKFMNS GQTCVAPDYI LCDPSIQNQI VEKLKKSLKE FYGEDAKKSR DYGRIISARH FQRVMGLIEG QKVAYGGTGD AATRYIAPTI LTDVDPQSPV MQEEIFGPVLPIVCVRSLEE AIQFINQREK PLALYMFSSN DKVIKKMIAE TSSGGVAAND VIVHITLHSL PFGGVGNSGM GSYHGKKSFE TFSHRRSCLV RPLMNDEGLK VRYPPSPAKM TQH | |
Greater than 95% as determined by SDS-PAGE. |
ALDH3A1 Protein | |
Research Use Only | |
ALDH3A1 | |
Human | |
His | |
ALDH3A1 solution containing 20mM Tris-HCl pH 8.0, 0.1M NaCl and 10% glycerol. |