missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Aldehyde Dehydrogenase 1-A1/ALDH1A1
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
Description
The Recombinant Human Aldehyde Dehydrogenase 1-A1/ALDH1A1 is derived from E. coli. The Recombinant Human Aldehyde Dehydrogenase 1-A1/ALDH1A1 has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in Tris, NaCl, DTT and Glycerol. |
| Gene ID (Entrez) | 216 |
| Molecular Weight (g/mol) | 56 kDa |
| Name | Aldehyde Dehydrogenase 1-A1/ALDH1A1 |
| Quantity | 20 μg |
| Source | E. coli-derived human Aldehyde Dehydrogenase 1-A1/ALDH1A1 protein Ser2-Ser501,with an N-terminal Met and 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. |
| Structural Form | Noncovalently-linked homotetramer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction