missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Alanine aminotransferase 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Alanine aminotransferase 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6114-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_005300.1 | |
Alanine aminotransferase 1 Protein | |
Research Use Only | |
GPT | |
Recombinant Protein | |
E. coli | |
ASSTGDRSQA VRHGLRAKVL TLDGMNPRVR RVEYAVRGPI VQRALELEQE LRQGVKKPFT EVIRANIGDA QAMGQRPITF LRQVLALCVN PDLLSSPNFP DDAKKRAERI LQACGGHSLG AYSVSSGIQL IREDVARYIE RRDGGIPADP NNVFLSTGAS DAIVTVLKLL VAGEGHTRTG VLIPIPQYPL YSATLAELGA VQVDYYLDEE RAWALDVAEL HRALGQARDH CRPRALCVIN PGNPTGQVQT RECIEAVIRF AFEERLFLLA DEVYQDNVYA AGSQFHSFKK VLMEMGPPYA GQQELASFHS TSKGYMGECG FRGGYVEVVN MDAAVQQQML KLMSVRLCPP VPGQALLDLV VSPPAPTDPS FAQFQAEKQA VLAELAAKAK LTEQVFNEAP GISCNPVQGA MYSFPRVQLP PRAVERAQEL GLAPDMFFCL RLLEETGICV VPGSGFGQRE GTYHFRMTIL PPLEKLRLLL EKLSRFHAKF TLEYS | |
Greater than 90% as determined by SDS-PAGE. |
2875 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |