missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human AKT3 His Protein

Product Code. 18326982
Change view
Click to view available options
Quantity:
10 μg
100 μg
20 μg
250 μg
50 μg
Unit Size:
100µg
10µg
20µg
250µg
50µg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18326982 20 μg 20µg
18358451 50 μg 50µg
18377014 250 μg 250µg
18398235 10 μg 10µg
18351787 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 18326982 Supplier Novus Biologicals™ Supplier No. NBP31399920UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.25 mg/ml
For Use With (Application) SDS-PAGE
Formulation PBS (pH 7.4), 10% glycerol
Gene ID (Entrez) 10000
Molecular Weight (g/mol) M.W. Theoretical: 56.5 kDa
Quantity 20 μg
Storage Requirements Store at 4C short term. Aliquot and Store at -20°C long term. Avoid freeze-thaw cycles.
Endotoxin Concentration <1 EU per 1ug of protein (determined by LAL method)
Gene Alias Akt-3, EC 2.7.11, EC 2.7.11.1, PKB gamma, PKBGDKFZp434N0250, PRKBG, v-akt murine thymoma viral oncogene homolog 3 (protein kinase B, gamma)
Gene Symbol AKT3
Research Category Cancer, Hypoxia, Protein Kinase, Signal Transduction
Purity or Quality Grade >90%, by SDS-PAGE
Protein AKT3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.