missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human AKR7A2 Protein

Product Code. 15904108
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
This item is not returnable. View return policy

Product Code. 15904108

Brand: enQuireBio™ QP105141mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the AKR7A2 was constructed and used to recombinantly synthesize the protein.

Specifications

Name AKR7A2 Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Biological Activity Specific activity: approximately 0.25-0.3 units/mg. Enzymatic activity was confirmed by measuring the amount of enzyme catalyzing the oxidation of 1 micromole NADPH per minute at 25C. Specific activity was expressed as units/mg protein.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSELEM LSAASRVVSR AAVHCALRSP PPEARALAMS RPPPPRVASV LGTMEMGRRM DAPASAAAVR AFLERGHTEL DTAFMYSDGQ SETILGGLGL GLGGGDCRVK IATKANPWDG KSLKPDSVRS QLETSLKRLQ CPQVDLFYLH APDHGTPVEE TLHACQRLHQ EGKFVELGLS NYASWEVAEI CTLCKSNGWI LPTVYQGMYN ATTRQVETEL FPCLRHFGLR FYAYNPLAGG LLTGKYKYED KDGKQPVGRF FGNSWAETYR NRFWKEHHFE AIALVEKALQ AAYGASAPSV TSAALRWMYH HSQLQGAHGD AVILGMSSLE QLEQNLAATE EGPLEPAVVD AFNQAWHLVA HECPNYFR
Buffer The AKR7A2 solution contains 20mM Tris-HCl pH 8.0, 1mM DTT and 20% glycerol.
Purity or Quality Grade Greater than 90.0% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.