missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AK2 / Adenylate kinase 2 Protein
A DNA sequence encoding the Homo sapiens (Human) AK2 / Adenylate kinase 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5635-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001186128.1 | |
AK2 / Adenylate kinase 2 Protein | |
Research Use Only | |
AK2 | |
Recombinant Protein | |
E. coli | |
MAPSVPAAEP EYPKGIRAVL LGPPGAGKGT QAPRLAENFC VCHLATGDML RAMVASGSEL GKKLKATMDA GKLVSDEMVV ELIEKNLETP LCKNGFLLDG FPRTVRQAEM LDDLMEKRKE KLDSVIEFSI PDSLLIRRIT GRLIHPKSGR SYHEEFNPPK EPMKDDITGE PLIRRSDDNE KALKIRLQAY HTQTTPLIEY YRKRGIHSAI DASQTPDVVF ASILAAFSKA TCKDLVMFI | |
Greater than 80% as determined by SDS-PAGE. |
204 | |
1 mg | |
Not determined. | |
Full Length of BC009405 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |