missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human AIP Protein

Product Code. 15975788
Change view
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15975788 5 μg 5µg
15965788 20 μg 20µg
15955788 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15975788 Supplier enQuireBio™ Supplier No. QP109885ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the AIP was constructed and used to recombinantly synthesize the protein.

Specifications

Name AIP Protein
Quantity 5 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence MGSSHHHHHH SSGLVPRGSH MADIIARLRE DGIQKRVIQE GRGELPDFQD GTKATFHYRT LHSDDEGTVL DDSRARGKPM ELIIGKKFKL PVWETIVCTM REGEIAQFLC DIKHVVLYPL VAKSLRNIAV GKDPLEGQRH CCGVAQMREH SSLGHADLDA LQQNPQPLIF HMEMLKVESP GTYQQDPWAM TDEEKAKAVP LIHQEGNRLY REGHVKEAAA KYYDAIACLK NLQMKEQPGS PEWIQLDKQI TPLLLNYCQC KLVVEEYYEV LDHCSSILNK YDDNVKAYFK RGKAHAAVWN AQEAQADFAK VLELDPALAP VVSRELRALE ARIRQKDEED KARFRGIFSH
Buffer AIP protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 1mM DTT and 10% glycerol.
Purity or Quality Grade Greater than 90.0% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.