missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human Agpat4 GST (N-Term) Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
2 μg
Packungsgröße:
2µg
Beschreibung
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Spezifikation
Spezifikation
| For Use With (Application) | Western Blot, ELISA, Protein Array, Immunoaffinity Purification |
| Formulation | 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
| Gene ID (Entrez) | 56895 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 70.4 kDa |
| Quantity | 2 μg |
| Storage Requirements | Store at -80°C. Avoid freeze-thaw cycles. |
| Gene Alias | 1-acylglycerol-3-phosphate O-acyltransferase 4 (lysophosphatidic acidacyltransferase, delta), 1-AGP acyltransferase 4,1-acylglycerol-3-phosphate O-acyltransferase 4,1-AGPAT 4, dJ473J16.2,1-acyl-sn-glycerol-3-phosphate acyltransferase delta, EC 2.3.1.51, LPAAT-delta1-AGPAT4, Lysophosphatidic acid acyltransferase delta, lysophosphatidic acid acyltransferase-delta (LPAAT-delta) |
| Gene Symbol | AGPAT4 |
| Purity or Quality Grade | >80% by SDS-PAGE and Coomassie blue staining |
| Protein | Agpat4 |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur