missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human aFGF / FGF1 Protein
Recombinant Protein
£80.80 - £1435.00
Specifications
Accession Number | NP_000791.1 |
---|---|
Gene ID (Entrez) | 2246 |
Name | aFGF / FGF1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15955929
|
enQuireBio™
QP5339-10UG |
10 μg |
£80.80
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15985929
|
enQuireBio™
QP5339-50UG |
50 μg |
£191.00
50µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15945929
|
enQuireBio™
QP5339-100UG |
100 μg |
£329.00
100µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15975929
|
enQuireBio™
QP5339-500UG |
500 μg |
£845.00
500µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15965929
|
enQuireBio™
QP5339-1MG |
1 mg |
£1435.00
1mg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_000791.1 | |
aFGF / FGF1 Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 2 ng/mL in the BALB/c 3T3 cell proliferation assay. The bioactivity acceptance criteria is set at 5.0 x 10^5 units/mg. | |
Human | |
Untagged | |
10 mM sodium phosphate, 75 mM sodium chloride, pH 7.5 |
2246 | |
Research Use Only | |
FGF1 | |
Recombinant Protein | |
E. coli | |
MFNLPPGNYK KPKLLYCSNG GHFLRILPDG TVDGTRDRSD QHIQLQLSAE SVGEVYIKST ETGQYLAMDT DGLLYGSQTP NEECLFLERL EENHYNTYIS KKHAEKNWFV GLKKNGSCKR GPRTHYGQKA ILFLPLPVSS D | |
Greater than 95% as determined by SDS-PAGE. |