missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human AFG3-like protein 1

A DNA sequence encoding the Homo sapiens (Human) AFG3-like protein 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8466-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16084761

  • £1735.00 / 1mg
Estimated Shipment: 11-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_006787.2
AFG3-like protein 1
Research Use Only
AFG3L2
Recombinant Protein
E. coli
ERVIGGLEKK TQVLQPEEKK TVAYHEAGHA VAGWYLEHAD PLLKVSIIPR GKGLGYAQYL PKEQYLYTKE QLLDRMCMTL GGRVSEEIFF GRITTGAQDD LRKVTQSAYA QIVQFGMNEK VGQISFDLPR QGDMVLEKPY SEATARLIDD EVRILINDAY KRTVALLTEK KADVEKVALL LLEKEVLDKN DMVELLGPRP FAEKSTYEEF
Greater than 90% as determined by SDS-PAGE.
10939
1 mg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers