missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ADP-ribosylation factor-like protein 1

A DNA sequence encoding the Homo sapiens (Human) ADP-ribosylation factor-like protein 6, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8213-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16003321

  • £384.00 / 100µg
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001265222.1
ADP-ribosylation factor-like protein 1
Research Use Only
ARL6
Recombinant Protein
E. coli
GLLDRLSVLL GLKKKEVHVL CLGLDNSGKT TIINKLKPSN AQSQNILPTI GFSIEKFKSS SLSFTVFDMS GQGRYRNLWE HYYKEGQAII FVIDSSDRLR MVVAKEELDT LLNHPDIKHR RIPILFFANK MDLRDAVTSV KVSQLLCLEN IKDKPWHICA SDAIKGEGLQ EGVDWLQDQI QTVKT
Greater than 80% as determined by SDS-PAGE.
84100
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers