missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ADP-ribosylation factor 4 Protein

A DNA sequence encoding the Homo sapiens (Human) ADP-ribosylation factor 4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8338-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16094091

  • £1735.00 / 1mg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001651.1
ADP-ribosylation factor 4 Protein
Research Use Only
ARF2
Recombinant Protein
E. coli
MGLTISSLFS RLFGKKQMRI LMVGLDAAGK TTILYKLKLG EIVTTIPTIG FNVETVEYKN ICFTVWDVGG QDRIRPLWKH YFQNTQGLIF VVDSNDRERI QEVADELQKM LLVDELRDAV LLLFANKQDL PNAMAISEMT DKLGLQSLRN RTWYVQATCA TQGTGLYEGL DWLSNELSKR
Greater than 90% as determined by SDS-PAGE.
378
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers