missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Adiponectin / Acrp30 / ADIPOQ Protein
Click to view available options
Quantity:
1 mg
100 μg
25 μg
5 μg
500 μg
Unit Size:
100µg
1mg
25µg
500µg
5µg
Specifications
Specifications
| Accession Number | NP_001171271.1 |
| Gene ID (Entrez) | 9370 |
| Name | Adiponectin / Acrp30 / ADIPOQ Protein |
| Quantity | 5 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Less than or equal to 5.00 EUs/ug as determined by the LAL method. |
| Gene Symbol | ADIPOQ |
| Biological Activity | gACRP (globular adiponectin) activity was confirmed by measuring inhibition of cytotoxicity of M1 cells. Typical ED50 values for our recombinant adiponectin in this assay are in the range of 3 ug/ml. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction