missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Adiponectin / Acrp30 / ADIPOQ Protein

Product Code. 15965829
Click to view available options
Quantity:
1 mg
100 μg
25 μg
5 μg
500 μg
Unit Size:
100µg
1mg
25µg
500µg
5µg
This item is not returnable. View return policy

Product Code. 15965829

Brand: enQuireBio™ QP52791mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Specifications

Accession Number NP_001171271.1
Gene ID (Entrez) 9370
Name Adiponectin / Acrp30 / ADIPOQ Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Less than or equal to 5.00 EUs/ug as determined by the LAL method.
Gene Symbol ADIPOQ
Biological Activity gACRP (globular adiponectin) activity was confirmed by measuring inhibition of cytotoxicity of M1 cells. Typical ED50 values for our recombinant adiponectin in this assay are in the range of 3 ug/ml.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag Untagged
Sequence MKGEPGEGAY VYRSAFSVGL ETYVTIPNMP IRFTKIFYNQ QNHYDGSTGK FHCNIPGLYY FAYHITVYMK DVKVSLFKKD KAMLFTYDQY QENNVDQASG SVLLHLEVGD QVWLQVYGEG ERNGLYADND NDSTFTGFLL YHDTN
Buffer 10 mM Tris, 0.75 mM DTT, pH 8.0
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.