missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ADAT1 Protein

A cDNA sequence encoding the ADAT1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10963-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15945708

  • £99.15 / 2µg
Estimated Shipment: 11-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

23536
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MWTADEIAQL CYEHYGIRLP KKGKPEPNHE WTLLAAVVKI QSPADKACDT PDKPVQVTKE VVSMGTGTKC IGQSKMRKNG DILNDSHAEV IARRSFQRYL LHQLQLAATL KEDSIFVPGT QKGVWKLRRD LIFVFFSSHT PCGDASIIPM LEFEDQPCCP VFRNWAHNSS VEASSNLEAP GNERKCEDPD SPVTKKMRLE PGTAAREVTN GAAHHQSFGK QKSGPISPGI HSCDLTVEGL ATVTRIAPGS AKVIDVYRTG AKCVPGEAGD SGKPGAAFHQ VGLLRVKPGR GDRTRSMSCS DKMARWNVLG CQGALLMHLL EEPIYLSAVV IGKCPYSQEA MQRALIGRCQ NVSALPKGFG VQELKILQSD LLFEQSRSAV QAKRADSPGR LVPCGAAISW SAVPEQPLDV TANGFPQGTT KKTIGSLQAR SQISKVELFR SFQKLLSRIA RDKWPHSLRV QKLDTYQEYK EAASSYQEAW STLRKQVFGS WIRNPPDYHQ FK
Greater than 95.0% as determined by SDS-PAGE.
ADAT1 Protein
Research Use Only
ADAT1
Human
Untagged
Adenosine Deaminase tRNA-Specific-1 at 0.1 mg/ml, 10mM Tris, pH 8.0, 0.1% Triton X-100, 0.002% NaN3.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers