missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ADAM15 / MDC15 Protein

A DNA sequence encoding the Homo sapiens (Human) ADAM15 / MDC15, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8390-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16064371

  • £384.00 / 100µg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001248393.1
ADAM15 / MDC15 Protein
Research Use Only
ADAM15
Recombinant Protein
E. coli
DVVTETKTVE LVIVADHSEA QKYRDFQHLL NRTLEVALLL DTFFRPLNVR VALVGLEAWT QRDLVEISPN PAVTLENFLH WRRAHLLPRL PHDSAQLVTG TSFSGPTVGM AIQNSICSPD FSGGVNMDHS TSILGVASSI AHELGHSLGL DHDLPGNSCP CPGPAPAKTC IMEASTDFLP GLNFSNCSRR ALEKALLDGM GSCLFERLPS LPPMAAFCGN MFVEPGEQCD CGFLDDCVDP CCDSLT
Greater than 90% as determined by SDS-PAGE.
8751
100 μg
Not determined.
Extracellular Domain
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers